Learn More
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158821000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-2-nitro-1, 3-propanediol | |
97.5 | |
>120.0°C | |
Authentic | |
Plastic bottle | |
HOCH2C(Br)(NO2)CH2OH | |
100 g | |
15, 1457 | |
Solubility in water: 250g/L at 22°C. Other solubilities: soluble in ethanol (500g/L at 24°C),glycerol,(10g/L at 23°C),propylene glycol (400g/L at,23°C),ethyl acetate;,slightly soluble in chcl3,acetone,ether,benzene | |
C(C(CO)([N+](=O)[O-])Br)O | |
199.99 | |
CHEBI:31306 | |
98% |
52-51-7 | |
100.0 | |
White to Yellow | |
97.5% min. (GC) | |
C3H6BrNO4 | |
MFCD00007390 | |
01, IV, 2501 | |
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol | |
LVDKZNITIUWNER-UHFFFAOYSA-N | |
2-bromo-2-nitropropane-1,3-diol | |
2450 | |
199.99 | |
Crystalline Powder or Crystals |
Chemical Identifiers
52-51-7 | |
199.99 | |
LVDKZNITIUWNER-UHFFFAOYSA-N | |
2450 | |
2-bromo-2-nitropropane-1,3-diol |
C3H6BrNO4 | |
MFCD00007390 | |
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol | |
CHEBI:31306 | |
C(C(CO)([N+](=O)[O-])Br)O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 200-143-
RUO – Research Use Only