missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-2-methylpropionic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 305200050

 View more versions of this product

Catalog No. AC305200050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5% (Acid)
41° to 46°C
5g
15, 1431
55527
Authentic
98%
C4H7BrO2
MFCD00004168
Liquid
167.01
Clear colorless to slightly yellow
100.0
Glass bottle
BrC(CH3)2CO2H
2-Bromo-2-methylpropionic acid
97.5
α-Bromoisobutyric acid

Chemical Identifiers

55527
100.0
C4H7BrO2
167.01
α-Bromoisobutyric acid
≥97.5% (Acid)
97.5
98%
BrC(CH3)2CO2H
MFCD00004168
2-Bromo-2-methylpropionic acid

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger