missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-2-methylpropane, 96%, stabilized, Acros Organics

Manufacturer:  Thermo Fisher Scientific 159265000

 View more versions of this product

Catalog No. AC159265000



Specifications

Specifications

2-Bromo-2-methylpropane
18°C
Glass bottle
1.18
72.0°C to 74.0°C
-20.0°C
507-19-7
C4H9Br
01,127
1-bromo-1,1-dimethylethane
RKSOPLXZQNSWAS-UHFFFAOYSA-N
2-bromo-2-methylpropane
10485
Liquid
1.1800g/mL
Authentic
1.4269 to 1.4289
Potassium carbonate
Undesignated
500mL
95% min. (GC)
(CH3)3CBr
15,1557
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
CC(C)(C)Br
137.02
137.02
96%

Chemical Identifiers

507-19-7
137.02
1-bromo-1,1-dimethylethane
2-bromo-2-methylpropane
C4H9Br
RKSOPLXZQNSWAS-UHFFFAOYSA-N
10485
CC(C)(C)Br
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 208-065-9

SDS Documents