missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-2'-chloroacetophenone, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427890010

 View more versions of this product

Catalog No. AC427890010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-2′-chloroacetophenone
>110°C
Consistent with structure
1.602
Colorless to Yellow
5000-66-8
100.0
C8H6BrClO
MFCD00832993
WZWWEVCLPKAQTA-UHFFFAOYSA-N
2-bromo-1-(2-chlorophenyl)ethanone
2735785
Liquid
1.6020g/mL
Authentic
Glass bottle
162°C to 166°C (20.0mmHg)
1g
94.0
94% min. (GC)
BrCH2C(O)C6H4Cl
2'-chloro-2-bromoacetophenone, 2'-chlorophenacyl bromide, 2-bromo-1-2-chlorophenyl ethan-1-one, 2-bromo-1-2-chlorophenyl ethanone, 2-bromo-1-2-chlorophenyl-1-ethanone, 2-bromo-2'-chloroacetophenone, 2-chlorophenacyl bromide, 2-chlorophenacylbromide, bromomethyl 2-chlorophenyl ketone, zlchem 501
C1=CC=C(C(=C1)C(=O)CBr)Cl
233.49
233.49
95%

Chemical Identifiers

5000-66-8
233.49
WZWWEVCLPKAQTA-UHFFFAOYSA-N
2735785
C1=CC=C(C(=C1)C(=O)CBr)Cl
C8H6BrClO
MFCD00832993
2'-chloro-2-bromoacetophenone, 2'-chlorophenacyl bromide, 2-bromo-1-2-chlorophenyl ethan-1-one, 2-bromo-1-2-chlorophenyl ethanone, 2-bromo-1-2-chlorophenyl-1-ethanone, 2-bromo-2'-chloroacetophenone, 2-chlorophenacyl bromide, 2-chlorophenacylbromide, bromomethyl 2-chlorophenyl ketone, zlchem 501
2-bromo-1-(2-chlorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents