Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-2'-chloroacetophenone, 95%

Chemical Identifiers

Chemical Name or Material 2-Bromo-2'-chloroacetophenone
CAS 5000-66-8
Molecular Formula C8H6BrClO
Formula Weight 233.5
MDL Number MFCD00832993
UN Number UN1760
Synonym 2-Chlorophenacyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2730303

  SDS
alfa aesar™
H2730303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2730306

  SDS
alfa aesar™
H2730306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.5
Chemical Name or Material 2-Bromo-2'-chloroacetophenone
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1760
Health Hazard 1 Danger
MDL Number MFCD00832993
Assay 95%
CAS 5000-66-8
Quantity 1g
Flash Point >110°C (230°F)
Molecular Formula C8H6BrClO
Recommended Storage 2° to 8°C
Boiling Point 105°C (1mmHg)
Synonym 2-Chlorophenacyl bromide
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C