Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-2-butene, cis + trans, 98%, stab.

Chemical Identifiers

Chemical Name or Material 2-Bromo-2-butene
CAS 13294-71-8
Name Note cis + trans, stabilized
Molecular Formula C4H7Br
Formula Weight 135
MDL Number MFCD00000141
UN Number UN1993
Beilstein 1733329
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1805814

  SDS
alfa aesar™
A1805814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1805822

  SDS
alfa aesar™
A1805822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135
Chemical Name or Material 2-Bromo-2-butene
TSCA TSCA
Name Note cis + trans, stabilized
Assay 98%
Molecular Formula C4H7Br
MDL Number MFCD00000141
CAS 13294-71-8
UN Number UN1993
Beilstein 1733329
Boiling Point 82° to 90°C
Quantity 25g
Flash Point 1°C (33°F)
Odor Recognizable
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Applications
Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.

Solubility
Not miscible or difficult to mix in water (664 mg/L 25 EST).

Notes
Store away from oxidizing agents and heat. Store away from electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.

2° to 8°C