Please login to your online account to display your discounted pricing

2-Bromo-2',5'-dimethoxyacetophenone 97%, Acros Organics


Quantity 10g

Manufacturer: thermo fisher scientific  147320100

 View more versions of this product

Catalog No. AC147320100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Fine Crystalline Powder or Needles
Formula Weight 259.09
Quantity 10g
Color Beige
EINECS Number 214-873-2
TSCA TSCA
Melting Point 83° to 88°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Bromo-2', 5'-dimethoxyacetophenone
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 97%
MDL Number MFCD00000198
CAS 1204-21-3
Linear Formula (CH3O)2C6H3COCH2Br
Molecular Formula C10H11BrO3
Name Note 97%
Health Hazard 1 LACHRYMATOR
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Bromo-2', 5'-dimethoxyacetophenone
Name Note 97%
CAS 1204-21-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H11BrO3
Linear Formula (CH3O)2C6H3COCH2Br
Formula Weight 259.09
MDL Number MFCD00000198
Accessories

LACHRYMATOR

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents