Please login to your online account to display your discounted pricing

2-Bromo-2',4'-dimethoxyacetophenone, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  190010010

 View more versions of this product

Catalog No. AC190010010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 259.09
Physical Form Fine Crystalline Powder
Quantity 1g
Color White to brown
TSCA TSCA
Melting Point 101° to 105°C
EINECS Number 262-542-6
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger
Name Note 98%
MDL Number MFCD00000197
Chemical Name or Material 2-Bromo-2', 4'-dimethoxyacetophenone
CAS Min % 97.5
Linear Formula (CH3O)2C6H3COCH2Br
Molecular Formula C10H11BrO3
CAS 60965-26-6
Assay Percent Range 98%
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Bromo-2', 4'-dimethoxyacetophenone
Name Note 98%
CAS 60965-26-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H11BrO3
Linear Formula (CH3O)2C6H3COCH2Br
Formula Weight 259.09
MDL Number MFCD00000197
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents