Please login to your online account to display your discounted pricing

2-Bromo-2',4'-dimethoxyacetophenone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-2', 4'-dimethoxyacetophenone
Name Note 98%
CAS 60965-26-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H11BrO3
Linear Formula (CH3O)2C6H3COCH2Br
Formula Weight 259.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC190010010

  SDS
thermo fisher scientific
190010010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Bromo-2', 4'-dimethoxyacetophenone
Name Note 98%
CAS 60965-26-6
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
EINECS Number 262-542-6
TSCA TSCA
Packaging Glass bottle
Physical Form Fine Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H11BrO3
Linear Formula (CH3O)2C6H3COCH2Br
Formula Weight 259.09
MDL Number MFCD00000197
Color White to brown
Melting Point 101° to 105°C
Quantity 1g
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.