missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-2',4'-difluoroacetophenone, 97%

Manufacturer:  Alfa Aesar™ H5644203

 View more versions of this product

Catalog No. AAH5644203



Specifications

Specifications

2-Bromo-2',4'-difluoroacetophenone
31°C to 35°C
102429-07-2
MFCD00142822
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
C1=CC(=C(C=C1F)F)C(=O)CBr
235.028
235.03
51°C (124°F)
1g
C8H5BrF2O
UN2925
CSGDTHXBRAAOHV-UHFFFAOYSA-N
2-bromo-1-(2,4-difluorophenyl)ethanone
2774185
97%

Chemical Identifiers

102429-07-2
235.028
CSGDTHXBRAAOHV-UHFFFAOYSA-N
2774185
C1=CC(=C(C=C1F)F)C(=O)CBr
C8H5BrF2O
MFCD00142822
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw
2-bromo-1-(2,4-difluorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H228-H314-H318-H302
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.

Documents