Learn More
2-Bromo-2',4'-difluoroacetophenone, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 442280050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Bromo-2', 4'-difluoroacetophenone | |
94.0 | |
51°C | |
94% min. (GC) | |
5g | |
CSGDTHXBRAAOHV-UHFFFAOYSA-N | |
2-bromo-1-(2,4-difluorophenyl)ethanone | |
2774185 | |
95% |
102429-07-2 | |
100.0 | |
Authentic | |
C8H5BrF2O | |
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw | |
C1=CC(=C(C=C1F)F)C(=O)CBr | |
235.03 | |
235.03 |
Chemical Identifiers
102429-07-2 | |
235.03 | |
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw | |
2-bromo-1-(2,4-difluorophenyl)ethanone |
C8H5BrF2O | |
CSGDTHXBRAAOHV-UHFFFAOYSA-N | |
2774185 | |
C1=CC(=C(C=C1F)F)C(=O)CBr |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call
GHS Signal Word: Danger
RUO â Research Use Only