Please login to your online account to display your discounted pricing

2-Bromo-2',4'-dichloroacetophenone, 97%, ACROS Organics™

$1,774.90 - $1,774.90

Chemical Identifiers

Chemical Name or Material 2-Bromo-2', 4'-dichloroacetophenone
Name Note 98%
CAS 2631-72-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5BrCl2O
Formula Weight 267.94
MDL Number MFCD00053005
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC344990250

  SDS
thermo fisher scientific
AC344990250
Glass bottle 25g Each for $1,774.90

AC344990050

  SDS
thermo fisher scientific
AC344990050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC344991000

  SDS
thermo fisher scientific
AC344991000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Low Melting Solid
Formula Weight 267.94
Quantity 25g
Color White to brown
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Name Note 98%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 2-Bromo-2', 4'-dichloroacetophenone
Assay Percent Range 97%
Molecular Formula C8H5BrCl2O
MDL Number MFCD00053005
CAS 2631-72-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.