Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-2',3',4',5',6'-pentafluoroacetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-2',3',4',5',6'-pentafluoroacetophenone
CAS 5122-16-7
Molecular Formula C8H2BrF5O
Formula Weight 289.01
MDL Number MFCD00484757
UN Number UN1759
Synonym 2,3,4,5,6-Pentafluorophenacyl bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2481903

  SDS
alfa aesar™
B2481903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2481906

  SDS
alfa aesar™
B2481906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2481914

  SDS
alfa aesar™
B2481914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 289.01
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1759
Melting Point 30° to 32°C
Synonym 2,3,4,5,6-Pentafluorophenacyl bromide
Chemical Name or Material 2-Bromo-2',3',4',5',6'-pentafluoroacetophenone
Assay 97%
CAS 5122-16-7
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C8H2BrF5O
Health Hazard 1 Danger
Quantity 1g
MDL Number MFCD00484757
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.