missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1H-benzimidazole, 97%, Maybridge

$216.55 - $216.55

Chemical Identifiers

CAS 54624-57-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5BrN2
Formula Weight 197.03
MDL Number MFCD00160009
Chemical Name or Material 2-Bromo-1H-benzimidazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC09010DA SDS Fisher Scientific Company
CC09010DA
Amber Glass Bottle 1g Each for $216.55
Specifications Description & Specifications

Chemical Identifiers

54624-57-6
100.0
C7H5BrN2
MFCD00160009
97%
96.5
97%
197.03
2-Bromo-1H-benzimidazole

Specifications

54624-57-6
100.0
1g
C7H5BrN2
MFCD00160009
97%
96.5
Amber Glass Bottle
97%
197.03
2-Bromo-1H-benzimidazole