Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-tritylimidazole, 97%

$491.70 - $491.70

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-tritylimidazole
CAS 67478-47-1
Molecular Formula C22H17BrN2
Formula Weight 389.29g/mol
MDL Number MFCD02179528
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5186906

 
alfa aesar™
H51869-06
Each for $491.70
Description & Specifications

Specifications

Melting Point 204° to 206°C
Quantity 5g
Chemical Name or Material 2-Bromo-1-tritylimidazole
CAS 67478-47-1
Assay 97%
Molecular Formula C22H17BrN2
Formula Weight 389.29g/mol
MDL Number MFCD02179528
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.