missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-phenylpropane, TCI America™

$242.92 - $843.34

Chemical Identifiers

CAS 2114-39-8
Molecular Formula C9H11Br
Molecular Weight (g/mol) 199.091
MDL Number MFCD00000149
InChI Key NVYOCAOZCSNIHR-UHFFFAOYSA-N
Synonym 2-bromopropyl benzene, 2-bromo-1-phenylpropane, 1-phenyl-2-bromopropane, 2-bromo-propyl-benzene, benzene, 2-bromopropyl, 2-bromanylpropylbenzene, 2-bromopropyl-benzene, 1-2-bromopropyl benzene, acmc-1cr50, 2-bromo-1-phenylpropane
PubChem CID 102754
IUPAC Name 2-bromopropylbenzene
SMILES CC(CC1=CC=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22905G View Documents TCI America
B22905G
5g Each for $242.92
 
B229025G View Documents TCI America
B229025G
25g Each for $843.34
 
Specifications

Chemical Identifiers

2114-39-8
199.091
NVYOCAOZCSNIHR-UHFFFAOYSA-N
102754
CC(CC1=CC=CC=C1)Br
C9H11Br
MFCD00000149
2-bromopropyl benzene, 2-bromo-1-phenylpropane, 1-phenyl-2-bromopropane, 2-bromo-propyl-benzene, benzene, 2-bromopropyl, 2-bromanylpropylbenzene, 2-bromopropyl-benzene, 1-2-bromopropyl benzene, acmc-1cr50, 2-bromo-1-phenylpropane
2-bromopropylbenzene

Specifications

Undesignated
2114-39-8
MFCD00000149
NVYOCAOZCSNIHR-UHFFFAOYSA-N
2-bromopropylbenzene
102754
Clear Liquid at 20°C
2-Bromo-1-phenylpropane
5g
C9H11Br
2-bromopropyl benzene, 2-bromo-1-phenylpropane, 1-phenyl-2-bromopropane, 2-bromo-propyl-benzene, benzene, 2-bromopropyl, 2-bromanylpropylbenzene, 2-bromopropyl-benzene, 1-2-bromopropyl benzene, acmc-1cr50, 2-bromo-1-phenylpropane
CC(CC1=CC=CC=C1)Br
199.091
199.09
≥93.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No