Please login to your online account to display your discounted pricing

2-bromo-1-phenyl-1H-imidazole, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  CC75910EA

 View more versions of this product

Catalog No. CC75910EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 10g
Chemical Name or Material 2-bromo-1-phenyl-1H-imidazole
CAS 103456605
Assay 97%
Molecular Formula C9H7BrN2
Formula Weight 223.07g/mol
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 2-bromo-1-phenyl-1H-imidazole
Assay 97%
Molecular Formula C9H7BrN2
Formula Weight 223.07g/mol
Accessories

GHS Signal Word: Warning

SDS & Safety Documents