missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-methylbenzimidazole, 97%

$68.62 - $1,084.11

Chemical Identifiers

CAS 49572-60-3
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.062
MDL Number MFCD00839528
InChI Key PROLKELCEPYEQT-UHFFFAOYSA-N
Synonym 2-bromo-1-methyl-1h-benzo d imidazole, 2-bromo-1-methyl-1h-benzoimidazole, 1h-benzimidazole, 2-bromo-1-methyl, 2-bromo-1-methyl-1h-benzimidazole, 2-bromo-1-methyl-1,3-benzodiazole, 1-methyl-2-bromo-1h-benzimidazole, 2-bromo-1-methyl-1h-1,3-benzodiazole
PubChem CID 797082
IUPAC Name 2-bromo-1-methylbenzimidazole
SMILES CN1C2=CC=CC=C2N=C1Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60075MD View Documents Alfa Aesar™
H60075MD
250mg Each for $68.62
 
AAH6007503 View Documents Alfa Aesar™
H6007503
1g Each for $243.65
 
AAH6007506 View Documents Alfa Aesar™
H6007506
5g Each for $1,084.11
 
Description

Description

Applications
It is a synthetic building block.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

49572-60-3
211.062
PROLKELCEPYEQT-UHFFFAOYSA-N
797082
CN1C2=CC=CC=C2N=C1Br
C8H7BrN2
MFCD00839528
2-bromo-1-methyl-1h-benzo d imidazole, 2-bromo-1-methyl-1h-benzoimidazole, 1h-benzimidazole, 2-bromo-1-methyl, 2-bromo-1-methyl-1h-benzimidazole, 2-bromo-1-methyl-1,3-benzodiazole, 1-methyl-2-bromo-1h-benzimidazole, 2-bromo-1-methyl-1h-1,3-benzodiazole
2-bromo-1-methylbenzimidazole

Specifications

250mg
C8H7BrN2
2-bromo-1-methyl-1h-benzo d imidazole, 2-bromo-1-methyl-1h-benzoimidazole, 1h-benzimidazole, 2-bromo-1-methyl, 2-bromo-1-methyl-1h-benzimidazole, 2-bromo-1-methyl-1,3-benzodiazole, 1-methyl-2-bromo-1h-benzimidazole, 2-bromo-1-methyl-1h-1,3-benzodiazole
CN1C2=CC=CC=C2N=C1Br
211.062
211.06
2-Bromo-1-methylbenzimidazole
49572-60-3
MFCD00839528
PROLKELCEPYEQT-UHFFFAOYSA-N
2-bromo-1-methylbenzimidazole
797082
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning