Please login to your online account to display your discounted pricing

2-Bromo-1-methyl-1H-imidazole, 95%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC435430010

 View more versions of this product

Catalog No. AC435430010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94.0%
Formula Weight 161
Quantity 1g
Color Clear colorless to pale yellow to pink
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Bromo-1-methyl-1H-imidazole
Assay Percent Range 95%
Molecular Formula C4H5BrN2
MDL Number MFCD02179525
CAS 16681-59-7
Health Hazard 2 GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye damage.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take off contaminated clothing and wash before reuse. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Specifications

Chemical Name or Material 2-Bromo-1-methyl-1H-imidazole
CAS 16681-59-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C4H5BrN2
Formula Weight 161
MDL Number MFCD02179525
Accessories

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take off contaminated clothing and wash before reuse. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye damage.

SDS & Safety Documents