missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-methyl-1H-imidazole, 95%, ACROS Organics™

Chemical Identifiers

CAS 16681-59-7
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C4H5BrN2
Formula Weight 161
MDL Number MFCD02179525
Chemical Name or Material 2-Bromo-1-methyl-1H-imidazole
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC435430010 SDS Thermo Fisher Scientific
AC435430010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

16681-59-7
100.0
C4H5BrN2
MFCD02179525
≥94.0%
94.0
95%
161
2-Bromo-1-methyl-1H-imidazole

Specifications

161
1g
94.0
Authentic
16681-59-7
95%
≥94.0%
Liquid
Clear colorless to pale yellow to pink
100.0
MFCD02179525
C4H5BrN2
2-Bromo-1-methyl-1H-imidazole

Safety and Handling

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take off contaminated clothing and wash before reuse. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.