Please login to your online account to display your discounted pricing

2-Bromo-1-methyl-1H-imidazole, 95%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC435430050

 View more versions of this product

Catalog No. AC435430050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 161
Physical Form Liquid
Assay ≥94.0%
Quantity 5g
Color Clear colorless to pale yellow to pink
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 2 GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye damage.
CAS 16681-59-7
MDL Number MFCD02179525
Assay Percent Range 95%
Chemical Name or Material 2-Bromo-1-methyl-1H-imidazole
Molecular Formula C4H5BrN2
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take off contaminated clothing and wash before reuse. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Specifications

Chemical Name or Material 2-Bromo-1-methyl-1H-imidazole
CAS 16681-59-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C4H5BrN2
Formula Weight 161
MDL Number MFCD02179525
Accessories

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Take off contaminated clothing and wash before reuse. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye damage.

SDS & Safety Documents