Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-indanone, tech. 85%

$52.43 - $200.41

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-indanone
CAS 1775-27-5
Purity Grade Technical Grade
Molecular Formula C9H7BrO
Formula Weight 211.06
MDL Number MFCD00041030
Beilstein 637789
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1245303

  SDS
alfa aesar™
L1245303
1g Each for $52.43

AAL1245306

  SDS
alfa aesar™
L1245306
5g Each for $200.41
Description & Specifications

Specifications

Chemical Name or Material 2-Bromo-1-indanone
CAS 1775-27-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
Assay 85%
Molecular Formula C9H7BrO
Formula Weight 211.06
MDL Number MFCD00041030
Melting Point 37° to 39°C
Beilstein 637789
Boiling Point 110° to 112°C (0.4mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.