missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-fluoro-4-nitrobenzene, 95%

Manufacturer:  Alfa Aesar™ H6404522

 View more versions of this product

Catalog No. AAH6404522



Description

Description

Applications
Reatant in the synthesis of N-fused tricyclic indoles, dimethyamine, benzofuran.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Bromo-1-fluoro-4-nitrobenzene
701-45-1
MFCD00055433
1-bromo-2-fluoro-5-nitrobenzene, 2-bromo-1-fluoro-4-nitro-benzene, 2-bromo-4-nitrofluorobenzene, 3-bromo-4-fluoro-1-nitrobenzene, 3-bromo-4-fluoronitrobenzene, 3-bromo-4-fluoronitrobezene, acmc-1birt, benzene, 2-bromo-1-fluoro-4-nitro, ksc494c7t, pubchem8499
FAWMTDSAMOCUAR-UHFFFAOYSA-N
2-bromo-1-fluoro-4-nitrobenzene
69699
Solid
100g
C6H3BrFNO2
2089367
Slightly soluble in water.
C1=CC(=C(C=C1[N+](=O)[O-])Br)F
219.997
220
95%

Chemical Identifiers

701-45-1
219.997
FAWMTDSAMOCUAR-UHFFFAOYSA-N
69699
C1=CC(=C(C=C1[N+](=O)[O-])Br)F
C6H3BrFNO2
MFCD00055433
1-bromo-2-fluoro-5-nitrobenzene, 2-bromo-1-fluoro-4-nitro-benzene, 2-bromo-4-nitrofluorobenzene, 3-bromo-4-fluoro-1-nitrobenzene, 3-bromo-4-fluoronitrobenzene, 3-bromo-4-fluoronitrobezene, acmc-1birt, benzene, 2-bromo-1-fluoro-4-nitro, ksc494c7t, pubchem8499
2-bromo-1-fluoro-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-855-6

TSCA : No

Recommended Storage : Ambient temperatures

Documents