Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-(ethoxymethyl)imidazole, 97%

$244.09 - $944.74

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-(ethoxymethyl)imidazole
CAS 850429-54-8
Molecular Formula C6H9BrN2O
Formula Weight 205.05
MDL Number MFCD04038915
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5186006

  SDS
alfa aesar™
H5186006
5g Each for $944.74

AAH5186003

  SDS
alfa aesar™
H5186003
1g Each for $244.09
Description & Specifications

Specifications

Formula Weight 205.05
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
MDL Number MFCD04038915
Molecular Formula C6H9BrN2O
Quantity 5g
CAS 850429-54-8
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Warning
Chemical Name or Material 2-Bromo-1-(ethoxymethyl)imidazole

Applications
2-Bromo-1-(ethoxymethyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Very slightly soluble in water (11 g/L at 25°C).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.