Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-chloro-4-fluorobenzene, 98+%

$86.19 - $358.61

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-chloro-4-fluorobenzene
CAS 201849-15-2
Molecular Formula C6H3BrClF
Formula Weight 209.45
MDL Number MFCD00061120
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2553214

  SDS
alfa aesar™
B2553214
25g Each for $358.61

AAB2553206

  SDS
alfa aesar™
B2553206
5g Each for $86.19
Description & Specifications

Specifications

Formula Weight 209.45
Quantity 25g
Molecular Formula C6H3BrClF
Assay ≥98%
MDL Number MFCD00061120
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Bromo-1-chloro-4-fluorobenzene
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 201849-15-2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.