missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-benzofuran, 97%, Maybridge

$144.84 - $144.84

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C8H5BrO
Formula Weight 197.03
Chemical Name or Material 2-Bromo-1-benzofuran
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC06610DA SDS Fisher Scientific Company
CC06610DA
1g Each for $144.84
CC06610EA SDS Fisher Scientific Company
CC06610EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
C8H5BrO
197.03
2-Bromo-1-benzofuran

Specifications

54008-77-4
1g
C8H5BrO
2-Bromo-1-benzofuran
Amber Glass Bottle
97%
197.03