missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-(5-phenyl-2-thienyl)-1-ethanone, 95%, Maybridge™

Manufacturer:  Fisher Scientific Company CC10505CB

 View more versions of this product

Catalog No. CC10505CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-1-(5-phenyl-2-thienyl)-1-ethanone
250mg
94.5
95%
MFCD02681997
BOIAFAJKOBVLGS-UHFFFAOYSA-N
2-bromo-1-(5-phenylthiophen-2-yl)ethanone
2776310
95%
Amber glass bottle
10531-43-8
100.0
C12H9BrOS
2-bromo-1-5-phenyl-2-thienyl-1-ethanone, 2-bromo-1-5-phenyl-2-thienyl-ethanone, 2-bromo-1-5-phenyl-2-thiophenyl ethanone, 2-bromo-1-5-phenylthien-2-yl ethan-1-one, 2-bromo-1-5-phenylthiophen-2-yl ethanone, 2-bromoacetyl-5-phenylthiophene, bromomethyl 5-phenyl-2-thienyl ketone, ethanone, 2-bromo-1-5-phenyl-2-thienyl, ethanone,2-bromo-1-5-phenyl-2-thienyl
C1=CC=C(C=C1)C2=CC=C(S2)C(=O)CBr
281.167
281.17

Chemical Identifiers

10531-43-8
281.167
BOIAFAJKOBVLGS-UHFFFAOYSA-N
2776310
C1=CC=C(C=C1)C2=CC=C(S2)C(=O)CBr
C12H9BrOS
MFCD02681997
2-bromo-1-5-phenyl-2-thienyl-1-ethanone, 2-bromo-1-5-phenyl-2-thienyl-ethanone, 2-bromo-1-5-phenyl-2-thiophenyl ethanone, 2-bromo-1-5-phenylthien-2-yl ethan-1-one, 2-bromo-1-5-phenylthiophen-2-yl ethanone, 2-bromoacetyl-5-phenylthiophene, bromomethyl 5-phenyl-2-thienyl ketone, ethanone, 2-bromo-1-5-phenyl-2-thienyl, ethanone,2-bromo-1-5-phenyl-2-thienyl
2-bromo-1-(5-phenylthiophen-2-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents