Please login to your online account to display your discounted pricing

2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone, ≥95%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC15505DA

 View more versions of this product

Catalog No. CC15505DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 279.14
Quantity 1g
Assay 95%
CAS Max % 100.0
Molecular Formula C12H11BrN2O
Assay Percent Range 95%
MDL Number MFCD02681921
CAS Min % 94.5
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber glass bottle
Chemical Name or Material 2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone
CAS 137577-00-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone
CAS 137577-00-5
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H11BrN2O
Formula Weight 279.14
MDL Number MFCD02681921
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents