Please login to your online account to display your discounted pricing

2-Bromo-1-(4-pentylphenyl)ethan-1-one, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB12841DA

 View more versions of this product

Catalog No. BTB12841DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 269.18
Quantity 1g
Assay 95%
CAS Max % 100.0
Molecular Formula C13H17BrO
Linear Formula CH3(CH2)4C6H4C(O)CH2Br
MDL Number MFCD00218837
CAS Min % 94.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Packaging Amber Glass Bottle
Chemical Name or Material 2-Bromo-1-(4-pentylphenyl)ethan-1-one
Assay Percent Range 95%
CAS 64328-68-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Bromo-1-(4-pentylphenyl)ethan-1-one
CAS 64328-68-3
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H17BrO
Linear Formula CH3(CH2)4C6H4C(O)CH2Br
Formula Weight 269.18
MDL Number MFCD00218837
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents