Please login to your online account to display your discounted pricing

2-Bromo-1-(4-methylphenyl)ethan-1-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-(4-methylphenyl)ethan-1-one
CAS 619-41-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9BrO
Linear Formula CH3C6H4COCH2Br
Formula Weight 213.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CD00282EA

 
fisher scientific company
CD00282EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CD00282EE

 
fisher scientific company
CD00282EE
Amber Glass Bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.07
Quantity 10g
Assay 97%
CAS Max % 100.0
Linear Formula CH3C6H4COCH2Br
Molecular Formula C9H9BrO
CAS 619-41-0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 2-Bromo-1-(4-methylphenyl)ethan-1-one
Assay Percent Range 97%
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.