Please login to your online account to display your discounted pricing

2-Bromo-1-(4-methylphenyl)ethan-1-one, 97%, Maybridge


Quantity 50g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD00282EE

 View more versions of this product

Catalog No. CD00282EE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 213.07
Quantity 50g
Packaging Amber Glass Bottle
Assay 97%
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 2-Bromo-1-(4-methylphenyl)ethan-1-one
Assay Percent Range 97%
Molecular Formula C9H9BrO
Linear Formula CH3C6H4COCH2Br
CAS 619-41-0
CAS Min % 96.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Bromo-1-(4-methylphenyl)ethan-1-one
CAS 619-41-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9BrO
Linear Formula CH3C6H4COCH2Br
Formula Weight 213.07
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents