Please login to your online account to display your discounted pricing

2-Bromo-1-(4-isobutylphenyl)propan-1-one, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RH01005DA

 View more versions of this product

Catalog No. RH01005DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 269.18
Quantity 1g
Assay 97%
CAS Min % 97.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3CH(Br)C(O)C6H4CH2CH(CH3)2
Chemical Name or Material 2-Bromo-1-(4-isobutylphenyl)propan-1-one
Assay Percent Range 97%
Molecular Formula C13H17BrO
Packaging Amber Glass Bottle
MDL Number MFCD00831700
CAS 80336-64-7
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 2-Bromo-1-(4-isobutylphenyl)propan-1-one
CAS 80336-64-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17BrO
Linear Formula CH3CH(Br)C(O)C6H4CH2CH(CH3)2
Formula Weight 269.18
MDL Number MFCD00831700
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents