missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1-(4-fluorophenyl)-1-propanone, 98%

$65.09 - $257.15

Chemical Identifiers

CAS 345-94-8
Molecular Formula C9H8BrFO
Molecular Weight (g/mol) 231.064
MDL Number MFCD11131405
InChI Key QKHHCXOSPQAQQI-UHFFFAOYSA-N
Synonym 1-4-fluorophenyl-2-bromo-1-propanone, 2-bromo-1-4-fluorophenyl propan-1-one, 2-bromo-1-4-fluorophenyl-1-propanone, 2-bromo-1-4-fluorophenyl-propan-1-one, 2-bromo-4'-fluoropropiophenone, 4-fluoro-alpha-bromopropiophenone, acmc-209i8k
PubChem CID 13051652
IUPAC Name 2-bromo-1-(4-fluorophenyl)propan-1-one
SMILES CC(C(=O)C1=CC=C(C=C1)F)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6133803
View Documents
Alfa Aesar™
H6133803
1g Each for $65.09
 
AAH6133806
View Documents
Alfa Aesar™
H6133806
5g Each for $257.15
 
Specifications

Chemical Identifiers

345-94-8
231.064
QKHHCXOSPQAQQI-UHFFFAOYSA-N
13051652
CC(C(=O)C1=CC=C(C=C1)F)Br
C9H8BrFO
MFCD11131405
1-4-fluorophenyl-2-bromo-1-propanone, 2-bromo-1-4-fluorophenyl propan-1-one, 2-bromo-1-4-fluorophenyl-1-propanone, 2-bromo-1-4-fluorophenyl-propan-1-one, 2-bromo-4'-fluoropropiophenone, 4-fluoro-alpha-bromopropiophenone, acmc-209i8k
2-bromo-1-(4-fluorophenyl)propan-1-one

Specifications

1g
C9H8BrFO
1-4-fluorophenyl-2-bromo-1-propanone, 2-bromo-1-4-fluorophenyl propan-1-one, 2-bromo-1-4-fluorophenyl-1-propanone, 2-bromo-1-4-fluorophenyl-propan-1-one, 2-bromo-4'-fluoropropiophenone, 4-fluoro-alpha-bromopropiophenone, acmc-209i8k
CC(C(=O)C1=CC=C(C=C1)F)Br
231.064
231.06
98%
345-94-8
MFCD11131405
QKHHCXOSPQAQQI-UHFFFAOYSA-N
2-bromo-1-(4-fluorophenyl)propan-1-one
13051652
Solid
2-Bromo-1-(4-fluorophenyl)-1-propanone
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures