Learn More
2-Bromo-1,4-dimethoxybenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1564018
Description
2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-1,4-dimethoxybenzene has been used in preparation of S-(-)-2-[N-(trifluoroacetyl)amino]-1-(2,5-dimethoxy-4- bromophenyl)-1-propanone, 1,4-bis{4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1,4-bis{2,5-dimethoxy-4-[2-(4-pyridyl)ethenyl]phenyl}butadiyne and 1-bromo-4-iodo-2,5-dimethoxybenzene. It can be used as a chemical redox shuttle additive in Lithium ion batteries for repeated overcharge and overdischarge protection.
Solubility
Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
Specifications
2-Bromo-1,4-dimethoxybenzene | |
1.441 | |
>110°C (230°F) | |
1.57 | |
50 g | |
1-bromo-2,5-dimethoxybenzene, benzene, 2-bromo-1,4-dimethoxy, 2,5-dimethoxybromobenzene, pubchem19785, 2-bromo-4-methoxyanisole, acmc-1cby3, ksc490m9p, 1,4-dimethoxy-2-bromobenzene, 1-bromo-2, 5-dimethoxybenzene, dwcgnrkflrlwcj-uhfffaoysa | |
DWCGNRKFLRLWCJ-UHFFFAOYSA-N | |
2-bromo-1,4-dimethoxybenzene | |
91353 | |
98% |
25245-34-5 | |
260°C to 262°C | |
C8H9BrO2 | |
MFCD00008355 | |
2441884 | |
Insoluble in water. | |
COC1=CC=C(OC)C(Br)=C1 | |
217.06 | |
217.07 |
Chemical Identifiers
25245-34-5 | |
217.06 | |
DWCGNRKFLRLWCJ-UHFFFAOYSA-N | |
2-bromo-1,4-dimethoxybenzene |
C8H9BrO2 | |
MFCD00008355 | |
91353 | |
COC1=CC=C(OC)C(Br)=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-756-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only