Please login to your online account to display your discounted pricing

2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7BrF2O2
Linear Formula CHF2OC6H4C(O)CH2Br
Formula Weight 265.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00461EA

  SDS
fisher scientific company
TL00461EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 265.05
Quantity 10g
Assay 97%
Linear Formula CHF2OC6H4C(O)CH2Br
Chemical Name or Material 2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one
Molecular Formula C9H7BrF2O2
CAS 141134-24-9
Assay Percent Range 97%