Please login to your online account to display your discounted pricing

2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  TL00461EA

 View more versions of this product

Catalog No. TL00461EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 265.05
Quantity 10g
Assay 97%
Chemical Name or Material 2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one
Assay Percent Range 97%
Molecular Formula C9H7BrF2O2
Linear Formula CHF2OC6H4C(O)CH2Br
CAS 141134-24-9

Specifications

Chemical Name or Material 2-Bromo-1-[4-(difluoromethoxy)phenyl]ethan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7BrF2O2
Linear Formula CHF2OC6H4C(O)CH2Br
Formula Weight 265.05