Please login to your online account to display your discounted pricing

2-Bromo-1-(4-chlorophenyl)-2-phenylethan-1-one, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Bromo-1-(4-chlorophenyl)-2-phenylethan-1-one
CAS 1889-78-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C14H10BrClO
Linear Formula C6H5CHBrC(O)C6H4Cl
Formula Weight 309.59
MDL Number MFCD00052201
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DP00542CB

 
fisher scientific company
DP00542CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.59
Quantity 250mg
Assay 95%
CAS Max % 100.0
Chemical Name or Material 2-Bromo-1-(4-chlorophenyl)-2-phenylethan-1-one
Linear Formula C6H5CHBrC(O)C6H4Cl
CAS 1889-78-7
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle
Assay Percent Range 95%
MDL Number MFCD00052201
CAS Min % 94.5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C14H10BrClO