missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1,4-benzoquinone 93.0+%, TCI America™
$333.46 - $1,219.37
Chemical Identifiers
CAS | 3958-82-5 |
---|---|
Molecular Formula | C6H3BrO2 |
Molecular Weight (g/mol) | 186.992 |
MDL Number | MFCD00189377 |
InChI Key | PYEQXZOIQJWYAH-UHFFFAOYSA-N |
Synonym | 2-Bromocyclohexadiene-1,4-dione, Bromoquinone |
PubChem CID | 151181 |
IUPAC Name | 2-bromocyclohexa-2,5-diene-1,4-dione |
SMILES | C1=CC(=O)C(=CC1=O)Br |
Chemical Identifiers
3958-82-5 | |
186.992 | |
PYEQXZOIQJWYAH-UHFFFAOYSA-N | |
151181 | |
C1=CC(=O)C(=CC1=O)Br |
C6H3BrO2 | |
MFCD00189377 | |
2-Bromocyclohexadiene-1,4-dione, Bromoquinone | |
2-bromocyclohexa-2,5-diene-1,4-dione |
Specifications
3958-82-5 | |
C6H3BrO2 | |
1 g | |
PYEQXZOIQJWYAH-UHFFFAOYSA-N | |
2-bromocyclohexa-2,5-diene-1,4-dione | |
151181 | |
≥93.0% (GC) | |
2-Bromo-1,4-benzoquinone |
55°C | |
MFCD00189377 | |
2-Bromocyclohexadiene-1,4-dione, Bromoquinone | |
C1=CC(=O)C(=CC1=O)Br | |
186.992 | |
186.99 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator