missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1-(3-thienyl)-1-ethanone, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC13105DE
Specifications
2-Bromo-1-(3-thienyl)-1-ethanone | |
97% | |
C6H5BrOS | |
5g | |
TXEJYUFJFSPCHH-UHFFFAOYSA-N | |
2-bromo-1-thiophen-3-ylethanone | |
2776379 | |
97% |
1468-82-2 | |
Amber Glass Bottle | |
MFCD02677695 | |
2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone | |
BrCC(=O)C1=CSC=C1 | |
205.07 | |
205.07 |
Chemical Identifiers
1468-82-2 | |
205.07 | |
TXEJYUFJFSPCHH-UHFFFAOYSA-N | |
2776379 | |
BrCC(=O)C1=CSC=C1 |
C6H5BrOS | |
MFCD02677695 | |
2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone | |
2-bromo-1-thiophen-3-ylethanone |