missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1,3-difluoro-5-iodobenzene 98.0+%, TCI America™
$107.84 - $428.02
Chemical Identifiers
CAS | 155906-10-8 |
---|---|
Molecular Formula | C6H2BrF2I |
Molecular Weight (g/mol) | 318.89 |
MDL Number | MFCD06797752 |
InChI Key | WOLGNRYEQPISPB-UHFFFAOYSA-N |
PubChem CID | 2761406 |
IUPAC Name | 2-bromo-1,3-difluoro-5-iodobenzene |
SMILES | FC1=CC(I)=CC(F)=C1Br |
Chemical Identifiers
155906-10-8 | |
318.89 | |
WOLGNRYEQPISPB-UHFFFAOYSA-N | |
2-bromo-1,3-difluoro-5-iodobenzene |
C6H2BrF2I | |
MFCD06797752 | |
2761406 | |
FC1=CC(I)=CC(F)=C1Br |
Specifications
155906-10-8 | |
C6H2BrF2I | |
5 g | |
FC1=CC(I)=CC(F)=C1Br | |
318.89 | |
318.89 | |
Crystalline Powder |
66°C | |
MFCD06797752 | |
WOLGNRYEQPISPB-UHFFFAOYSA-N | |
2-bromo-1,3-difluoro-5-iodobenzene | |
2761406 | |
≥98.0% (GC) | |
2-Bromo-1,3-difluoro-5-iodobenzene |
Safety and Handling
TSCA : No