Please login to your online account to display your discounted pricing

2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one, Tech., Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB09098DA

 View more versions of this product

Catalog No. BTB09098DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 261.55
Quantity 1g
Assay Tech.
CAS Min % 80.0
CAS Max % 100.0
Chemical Name or Material 2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one
CAS 175135-93-0
Packaging Amber Glass Bottle
Molecular Formula C10H10BrClO
MDL Number MFCD00067862
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one
CAS 175135-93-0
CAS Min % 80.0
CAS Max % 100.0
Assay Tech.
Molecular Formula C10H10BrClO
Formula Weight 261.55
MDL Number MFCD00067862
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents