missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1,3-benzothiazole, 95%, Maybridge

Manufacturer:  Fisher Scientific Company CC06010DA

 View more versions of this product

Catalog No. CC06010DA


 

Specifications Description & Specifications

Specifications

214.09
97%
Amber Glass Bottle
2516-40-7
2-Bromo-1, 3-benzothiazole
MFCD02681887
1g
100.0
C7H4BrNS
95%
96.5

Chemical Identifiers

2516-40-7
100.0
C7H4BrNS
MFCD02681887
97%
96.5
95%
214.09
2-Bromo-1, 3-benzothiazole

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.