missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1,3,5-triisopropylbenzene, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1569236

 View more versions of this product

Catalog No. AAA1569236



Description

Description

2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Bromo-1,3,5-triisopropylbenzene
125°C to 126°C (4 mmHg)
1.5225
500 g
2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf
FUMMYHVKFAHQST-UHFFFAOYSA-N
2-bromo-1,3,5-tri(propan-2-yl)benzene
140846
96%
21524-34-5
C15H23Br
MFCD00051547
1953440
Slightly miscible with water.
CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C
283.253
283.25

Chemical Identifiers

21524-34-5
283.253
FUMMYHVKFAHQST-UHFFFAOYSA-N
140846
CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C
C15H23Br
MFCD00051547
2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf
2-bromo-1,3,5-tri(propan-2-yl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only