Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-1,3,4-trifluorobenzene, 97%

$232.43 - $232.43

Chemical Identifiers

Chemical Name or Material 2-Bromo-1,3,4-trifluorobenzene
CAS 176793-04-7
Molecular Formula C6H2BrF3
Formula Weight 210.98
MDL Number MFCD00236214
Synonym 2,3,6-Trifluorobromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2020309

 
alfa aesar™
B2020309
10g Each for $232.43
Description & Specifications

Specifications

Formula Weight 210.98
Chemical Name or Material 2-Bromo-1,3,4-trifluorobenzene
Assay 97%
Molecular Formula C6H2BrF3
MDL Number MFCD00236214
CAS 176793-04-7
Synonym 2,3,6-Trifluorobromobenzene
Quantity 10g
Flash Point 62°C (143°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.