missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1,3,4-thiadiazole, 97%, Thermo Scientific™

$157.67 - $157.67

Chemical Identifiers

CAS 61929-24-6
Molecular Formula C2HBrN2S
Molecular Weight (g/mol) 165.008
MDL Number MFCD04115375
InChI Key DQBCRVIBTFHJLM-UHFFFAOYSA-N
Synonym sftdmqbhhryszzp@, 2-bromo 1,3,4-thiadiazole, 2-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazole,2-bromo, 2-bromanyl-1,3,4-thiadiazole, 1,3,4-thiadiazole, 2-bromo, 2-bromo-1,3,4-thiadiazole
PubChem CID 2795198
IUPAC Name 2-bromo-1,3,4-thiadiazole
SMILES C1=NN=C(S1)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC08110DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC08110DA
1 g Amber Glass Bottle
Each for $157.67
 
Description

Description

Specifications

Chemical Identifiers

61929-24-6
165.008
DQBCRVIBTFHJLM-UHFFFAOYSA-N
2795198
C1=NN=C(S1)Br
C2HBrN2S
MFCD04115375
sftdmqbhhryszzp@, 2-bromo 1,3,4-thiadiazole, 2-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazole,2-bromo, 2-bromanyl-1,3,4-thiadiazole, 1,3,4-thiadiazole, 2-bromo, 2-bromo-1,3,4-thiadiazole
2-bromo-1,3,4-thiadiazole

Specifications

61929-24-6
100.0
C2HBrN2S
Amber Glass Bottle
sftdmqbhhryszzp@, 2-bromo 1,3,4-thiadiazole, 2-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazole,2-bromo, 2-bromanyl-1,3,4-thiadiazole, 1,3,4-thiadiazole, 2-bromo, 2-bromo-1,3,4-thiadiazole
C1=NN=C(S1)Br
165.008
165.01
2-Bromo-1, 3, 4-thiadiazole
96.5
97%
MFCD04115375
1 g
DQBCRVIBTFHJLM-UHFFFAOYSA-N
2-bromo-1,3,4-thiadiazole
2795198
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS