missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CD00248DA
Specifications
2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one | |
97% | |
C10H11BrO | |
1g | |
GSCCVWPVPFIRJP-UHFFFAOYSA-N | |
2-bromo-1-(2,4-dimethylphenyl)ethanone | |
2063450 | |
97% |
26346-85-0 | |
Amber Glass Bottle | |
(CH3)2C6H3C(O)CH2Br | |
2-bromo-1-2,4-dimethylphenyl ethanone, 2-bromo-1-2,4-dimethylphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethylphenyl, 2-bromo-2',4'-dimethylacetophenone, 2,4-dimethylphenacyl bromide, 2,4-dimethyl phenacyl bromide, pubchem16796, 2,4-dimethylphenacyl bromid, ksc557o1l, alpha-bromo-2',4'-dimethylacetophenone | |
CC1=CC(=C(C=C1)C(=O)CBr)C | |
227.101 | |
227.1 |
Chemical Identifiers
26346-85-0 | |
227.101 | |
2-bromo-1-2,4-dimethylphenyl ethanone, 2-bromo-1-2,4-dimethylphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethylphenyl, 2-bromo-2',4'-dimethylacetophenone, 2,4-dimethylphenacyl bromide, 2,4-dimethyl phenacyl bromide, pubchem16796, 2,4-dimethylphenacyl bromid, ksc557o1l, alpha-bromo-2',4'-dimethylacetophenone | |
2-bromo-1-(2,4-dimethylphenyl)ethanone |
C10H11BrO | |
GSCCVWPVPFIRJP-UHFFFAOYSA-N | |
2063450 | |
CC1=CC(=C(C=C1)C(=O)CBr)C |