Please login to your online account to display your discounted pricing

2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, Maybridge


Quantity 25g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD00248EB

 View more versions of this product

Catalog No. CD00248EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 227.1
Quantity 25g
Assay 97%
Chemical Name or Material 2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H11BrO
Linear Formula (CH3)2C6H3C(O)CH2Br
CAS 26346-85-0

Specifications

Chemical Name or Material 2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11BrO
Linear Formula (CH3)2C6H3C(O)CH2Br
Formula Weight 227.1