Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)pyridine-5-carboxaldehyde, 97%

$104.53 - $360.19

Chemical Identifiers

Chemical Name or Material 2-(Boc-amino)pyridine-5-carboxaldehyde
CAS 199296-40-7
Molecular Formula C11H14N2O3
Formula Weight 222.25
MDL Number MFCD08064228
Synonym tert-Butyl 5-formyl-2-pyridylcarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5007503

 
alfa aesar™
H5007503
1g Each for $360.19

AAH50075MD

 
alfa aesar™
H50075MD
250mg Each for $104.53
Description & Specifications

Specifications

Formula Weight 222.25
Chemical Name or Material 2-(Boc-amino)pyridine-5-carboxaldehyde
Assay 97%
Molecular Formula C11H14N2O3
MDL Number MFCD08064228
Quantity 1g
CAS 199296-40-7
Synonym tert-Butyl 5-formyl-2-pyridylcarbamate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

Solubility
Soluble in water.

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.