missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)ethyl bromide, 96%

Manufacturer:  Alfa Aesar™ H5521103

 View more versions of this product

Catalog No. AAH5521103



Description

Description

Applications
N-t-Boc-2-bromoethylamine is a building block for preparing fluorinated spacers having nucleophilic and electrophilic termini.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store at -20°C. Reactive with oxidizing agents.
Specifications

Specifications

2-(Boc-amino)ethyl bromide
1g
C7H14BrNO2
MFCD02683428
tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino ethyl bromide, 2-boc-amino-ethyl bromide
CC(C)(C)OC(=O)NCCBr
224.098
224.1
30°C to 32°C
39684-80-5
BrCH2CH2NHCO2C(CH3)3
2325117
TZRQZPMQUXEZMC-UHFFFAOYSA-N
tert-butyl N-(2-bromoethyl)carbamate
4103526
96%

Chemical Identifiers

39684-80-5
224.098
TZRQZPMQUXEZMC-UHFFFAOYSA-N
4103526
CC(C)(C)OC(=O)NCCBr
C7H14BrNO2
MFCD02683428
tert-butyl n-2-bromoethyl carbamate, 2-boc-amino ethyl bromide, n-boc-bromoethylamine, tert-butyl 2-bromoethyl carbamate, n-boc-2-bromoethylamine, tert-butyl 2-bromoethylcarbamate, 2-bromo-ethyl-carbamic acid tert-butyl ester, 2-tert-butoxycarbonylamino ethyl bromide, 2-boc-amino ethyl bromide, 2-boc-amino-ethyl bromide
tert-butyl N-(2-bromoethyl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : −30° to −10°C

SDS Documents