Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Boc-amino)-5-pyridinemethanol, 97%

$281.77 - $1,241.57

Chemical Identifiers

Chemical Name or Material 2-(Boc-amino)-5-pyridinemethanol
CAS 169280-83-5
Molecular Formula C11H16N2O3
Formula Weight 224.26
MDL Number MFCD07186264
Synonym tert-Butyl (5-hydroxymethyl-2-pyridyl)carbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5007703

  SDS
alfa aesar™
H5007703
1g Each for $281.77

AAH5007706

  SDS
alfa aesar™
H5007706
5g Each for $1,241.57
Description & Specifications

Specifications

Formula Weight 224.26
Chemical Name or Material 2-(Boc-amino)-5-pyridinemethanol
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD07186264
CAS 169280-83-5
Molecular Formula C11H16N2O3
Synonym tert-Butyl (5-hydroxymethyl-2-pyridyl)carbamate
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.